siRNA Details
| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107224.2 | NP_001100694.1 | 958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk32b| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107224.2 | NP_001100694.1 | 958 | |||