siRNA Details
| Gene Symbol: Sorcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107225.2 | NP_001100695.2 | 17 | 2329 | ||
| XM_008770280.1 | XP_008768502.1 | 17 | 1800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sorcs2| Gene Symbol: Sorcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107225.2 | NP_001100695.2 | 17 | 2329 | ||
| XM_008770280.1 | XP_008768502.1 | 17 | 1800 | |||