siRNA Details
Gene Symbol: Trmt44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130061.1 | NP_001123533.1 | 4 | 839 | ||
GenBank mRNA | FQ212309.1 | - | 4 | 819 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trmt44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130061.1 | NP_001123533.1 | 4 | 839 | ||
GenBank mRNA | FQ212309.1 | - | 4 | 819 |