siRNA Details
| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130062.1 | NP_001123534.1 | 6 | 742 | ||
| XM_006251239.2 | XP_006251301.1 | 7 | 1559 | |||
| XM_008770283.1 | XP_008768505.1 | 7 | 1559 | |||
| GenBank mRNA | FQ223777.1 | - | 7 | 861 | ||

Dok7
Genomic Map