siRNA Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191680.1 | NP_001178609.1 | 2 | 246 | ||
| XM_006251243.2 | XP_006251305.1 | 3 | 548 | |||
| XM_006251244.2 | XP_006251306.1 | 3 | 545 | |||
| XM_006251245.2 | XP_006251307.1 | 3 | 381 | |||
| XM_006251246.2 | XP_006251308.1 | 3 | 378 | |||

Mfsd10
Genomic Map