siRNA Details
| Gene Symbol: Sh3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100684.1 | NP_001094154.1 | 2 | 235 | ||
| XM_006251247.2 | XP_006251309.1 | 2 | 1274 | |||
| XM_006251248.2 | XP_006251310.1 | 2 | 1112 | |||
| XM_006251249.2 | XP_006251311.1 | 2 | 1326 | |||
| XM_006251251.2 | XP_006251313.1 | 2 | 184 | |||
| XM_006251252.2 | XP_006251314.1 | 2 | 3812 | |||
| XM_006251253.2 | XP_006251315.1 | 2 | 1048 | |||
| XM_008770284.1 | XP_008768506.1 | 3 | 4779 | |||
| XM_008770285.1 | XP_008768507.1 | 2 | 441 | |||
| GenBank mRNA | CB762696.1 | - | 2 | 243 | ||
| FQ218289.1 | - | 2 | 183 | |||

Sh3bp2
Genomic Map