siRNA Details
Gene Symbol: Fam193a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_223536.8 | XP_223536.5 | 16 | 3008 |
Gene Symbol: LOC103690085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251409.2 | XP_006251471.1 | 3 | 1948 |

More Information
Other Species Targeted

