siRNA Details
| Gene Symbol: Cfap99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770377.1 | XP_008768599.1 | 6 | 696 | ||
| XM_008770378.1 | XP_008768600.1 | 7 | 807 | |||
| XM_008770379.1 | XP_008768601.1 | 6 | 719 | |||
| XM_008770380.1 | XP_008768602.1 | 4 | 4376 | |||
| XM_008770381.1 | XP_008768603.1 | 5 | 1384 | |||

Cfap99
Genomic Map