siRNA Details
| Gene Symbol: Cfap99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770377.1 | XP_008768599.1 | 9 | 973 | ||
| XM_008770378.1 | XP_008768600.1 | 10 | 1084 | |||
| XM_008770379.1 | XP_008768601.1 | 9 | 996 | |||
| XM_008770380.1 | XP_008768602.1 | 7 | 4653 | |||
| XM_008770381.1 | XP_008768603.1 | 8 | 1661 | |||

Cfap99
Genomic Map