siRNA Details
| Gene Symbol: Letm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005884.1 | NP_001005884.1 | 13, 14 | 2262 | ||
| GenBank mRNA | BC083642.1 | - | 13, 14 | 2262 | 7134917 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Letm1| Gene Symbol: Letm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005884.1 | NP_001005884.1 | 13, 14 | 2262 | ||
| GenBank mRNA | BC083642.1 | - | 13, 14 | 2262 | 7134917 | |