siRNA Details
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005884.1 | NP_001005884.1 | 12 | 1977 | ||
GenBank mRNA | BC083642.1 | - | 12 | 1977 | 7134917 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005884.1 | NP_001005884.1 | 12 | 1977 | ||
GenBank mRNA | BC083642.1 | - | 12 | 1977 | 7134917 |