siRNA Details
Gene Symbol: Tmem129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126274.1 | NP_001119746.1 | 4 | 1929 | ||
GenBank mRNA | BC099194.1 | - | 3 | 1397 | ||
BC161857.1 | - | 4 | 1929 | 9023916 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126274.1 | NP_001119746.1 | 4 | 1929 | ||
GenBank mRNA | BC099194.1 | - | 3 | 1397 | ||
BC161857.1 | - | 4 | 1929 | 9023916 |