siRNA Details
| Gene Symbol: Tmem129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126274.1 | NP_001119746.1 | 2 | 503 | ||
| GenBank mRNA | BC161857.1 | - | 2 | 503 | 9023916 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem129| Gene Symbol: Tmem129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126274.1 | NP_001119746.1 | 2 | 503 | ||
| GenBank mRNA | BC161857.1 | - | 2 | 503 | 9023916 | |