siRNA Details
| Gene Symbol: Fam53a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107228.2 | NP_001100698.1 | 6 | 1625 | ||
| XM_006251262.2 | XP_006251324.1 | 6 | 1686 | |||
| XM_006251266.2 | XP_006251328.1 | 5 | 1587 | |||
| XM_006251267.2 | XP_006251329.1 | 5 | 1635 | |||
| XM_006251268.2 | XP_006251330.1 | 5 | 1560 | |||
| XM_006251270.2 | XP_006251332.1 | 5 | 1100 | |||
| XM_008770286.1 | XP_008768508.1 | 5 | 2073 | |||
| XM_008770287.1 | XP_008768509.1 | 5 | 2077 | |||
| XM_008770288.1 | XP_008768510.1 | 5 | 1104 | |||
| GenBank mRNA | BC168895.1 | - | 6 | 1625 | 7312953 | |

Fam53a
Genomic Map