siRNA Details
| Gene Symbol: Depdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107229.1 | NP_001100699.1 | 40 | 4431 | ||
| XM_006251277.2 | XP_006251339.1 | 42 | 4696 | |||
| XM_006251279.2 | XP_006251341.1 | 42 | 4669 | |||
| XM_006251281.2 | XP_006251343.1 | 41 | 4603 | |||
| XM_008770289.1 | XP_008768511.1 | 41 | 4534 | |||
| XM_008770290.1 | XP_008768512.1 | 42 | 4668 | |||
| XM_008770291.1 | XP_008768513.1 | 42 | 4652 | |||
| XM_008770292.1 | XP_008768514.1 | 41 | 4522 | |||
| XM_008770293.1 | XP_008768515.1 | 41 | 4630 | |||

Depdc5
Genomic Map