siRNA Details
| Gene Symbol: Patz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107231.2 | NP_001100701.1 | 1 | 1122 | ||
| NM_001277214.1 | NP_001264143.1 | 1 | 1122 | |||
| NM_001277215.1 | NP_001264144.1 | 1 | 1122 | |||
| XM_006251292.2 | XP_006251354.1 | 1 | 1144 | |||

Patz1
Genomic Map