siRNA Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107232.1 | NP_001100702.1 | 12 | 2222 | ||
| XM_008770297.1 | XP_008768519.1 | 12 | 2255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbp2| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107232.1 | NP_001100702.1 | 12 | 2222 | ||
| XM_008770297.1 | XP_008768519.1 | 12 | 2255 | |||