siRNA Details
| Gene Symbol: Sf3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107235.1 | NP_001100705.1 | 9, 10 | 1504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3a1| Gene Symbol: Sf3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107235.1 | NP_001100705.1 | 9, 10 | 1504 | ||