siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077660.2 | NP_001071128.1 | 6 | 2471 | ||
| XM_006251312.2 | XP_006251374.1 | 6 | 2932 | |||
| XM_006251313.2 | XP_006251375.1 | 6 | 2761 | |||
| XM_006251315.2 | XP_006251377.1 | 6 | 2564 | |||
| XM_008770307.1 | XP_008768529.1 | 5 | 2453 | |||
| GenBank mRNA | EX490902.1 | - | 1 | 537 | ||
| FQ209875.1 | - | 6 | 2491 | |||
| FQ222248.1 | - | 6 | 2463 | |||
| FQ223767.1 | - | 6 | 2458 | |||

Urgcp
Genomic Map