siRNA Details
| Gene Symbol: Cobl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107236.1 | NP_001100706.1 | 8 | 2846 | ||
| XM_008770308.1 | XP_008768530.1 | 10 | 4532 | |||
| XM_008770309.1 | XP_008768531.1 | 9 | 4457 | |||
| XM_008770311.1 | XP_008768533.1 | 10 | 4629 | |||

Cobl
Genomic Map