siRNA Details
| Gene Symbol: Tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107238.1 | NP_001100708.1 | 3 | 663 | ||
| XM_006251444.2 | XP_006251506.1 | 4 | 501 | |||
| XM_006251445.2 | XP_006251507.1 | 4 | 502 | |||
| GenBank mRNA | BC168726.1 | - | 4 | 473 | 6889842 | |
| BC168735.1 | - | 4 | 473 | 6891034 | ||
| BC168766.1 | - | 4 | 473 | 6890852 | ||

Tmed4
Genomic Map