siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251532.2 | XP_006251594.1 | 7, 8 | 2664 | ||
| XM_006251534.2 | XP_006251596.1 | 7, 8 | 2501 | |||
| XM_008770443.1 | XP_008768665.1 | 8, 9 | 2818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aftph| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251532.2 | XP_006251594.1 | 7, 8 | 2664 | ||
| XM_006251534.2 | XP_006251596.1 | 7, 8 | 2501 | |||
| XM_008770443.1 | XP_008768665.1 | 8, 9 | 2818 | |||