siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251532.2 | XP_006251594.1 | 2, 3 | 2156 | ||
| XM_006251534.2 | XP_006251596.1 | 2, 3 | 1993 | |||
| XM_008770443.1 | XP_008768665.1 | 3, 4 | 2310 | |||
| XM_223651.8 | XP_223651.5 | 2, 3 | 2155 | |||
| GenBank mRNA | FQ230354.1 | - | 1, 2 | 346 | ||

Aftph
Genomic Map