siRNA Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100565.1 | NP_001094035.1 | 5 | 756 | ||
| GenBank mRNA | BC166745.1 | - | 5 | 589 | 7939059 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peli1| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100565.1 | NP_001094035.1 | 5 | 756 | ||
| GenBank mRNA | BC166745.1 | - | 5 | 589 | 7939059 | |