siRNA Details
| Gene Symbol: Wdpcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127537.1 | NP_001121009.1 | 4, 5 | 449 | ||
| GenBank mRNA | BC161962.1 | - | 4, 5 | 449 | 9027983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdpcp| Gene Symbol: Wdpcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127537.1 | NP_001121009.1 | 4, 5 | 449 | ||
| GenBank mRNA | BC161962.1 | - | 4, 5 | 449 | 9027983 | |