siRNA Details
| Gene Symbol: Ahsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107241.1 | NP_001100711.1 | 9 | 964 | ||
| XM_006251539.2 | XP_006251601.1 | 10 | 1099 | |||
| XM_006251540.2 | XP_006251602.1 | 9 | 987 | |||
| XM_006251541.2 | XP_006251603.1 | 9 | 981 | |||
| XM_006251542.2 | XP_006251604.1 | 9 | 1014 | |||
| GenBank mRNA | BC099114.1 | - | 10 | 1003 | ||

Ahsa2
Genomic Map