siRNA Details
| Gene Symbol: RGD1305110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100971.1 | NP_001094441.1 | 6 | 782 | ||
| XM_006251544.2 | XP_006251606.1 | 6 | 781 | |||
| GenBank mRNA | BC128776.1 | - | 6 | 782 | 7133856 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305110| Gene Symbol: RGD1305110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100971.1 | NP_001094441.1 | 6 | 782 | ||
| XM_006251544.2 | XP_006251606.1 | 6 | 781 | |||
| GenBank mRNA | BC128776.1 | - | 6 | 782 | 7133856 | |