siRNA Details
| Gene Symbol: Pex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107242.2 | NP_001100712.1 | 3 | 936 | ||
| GenBank mRNA | FM088835.1 | - | 2 | 585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex13| Gene Symbol: Pex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107242.2 | NP_001100712.1 | 3 | 936 | ||
| GenBank mRNA | FM088835.1 | - | 2 | 585 | ||