siRNA Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191684.1 | NP_001178613.1 | 5 | 342 | ||
| XM_006251597.2 | XP_006251659.1 | 5 | 1110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fancl| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191684.1 | NP_001178613.1 | 5 | 342 | ||
| XM_006251597.2 | XP_006251659.1 | 5 | 1110 | |||