siRNA Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013991.1 | NP_001014013.1 | 2 | 199 | ||
| XM_006250558.2 | XP_006250620.1 | 2 | 655 | |||
| XM_008769936.1 | XP_008768158.1 | 1 | 1047 | |||
| GenBank mRNA | BC079245.1 | - | 2 | 199 | 7132812 | |

Tmem175
Genomic Map