siRNA Details
Gene Symbol: Synpo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770581.1 | XP_008768803.1 | 2 | 2008 | ||
XM_223791.8 | XP_223791.6 | 4 | 2366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Synpo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770581.1 | XP_008768803.1 | 2 | 2008 | ||
XM_223791.8 | XP_223791.6 | 4 | 2366 |