siRNA Details
| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107248.1 | NP_001100718.1 | 15 | 2201 | ||
| XM_006251639.2 | XP_006251701.1 | 15 | 2365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcl| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107248.1 | NP_001100718.1 | 15 | 2201 | ||
| XM_006251639.2 | XP_006251701.1 | 15 | 2365 | |||