siRNA Details
| Gene Symbol: Zfp503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107250.1 | NP_001100720.1 | 2 | 975 | ||
| GenBank mRNA | BC166993.1 | - | 2 | 990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp503| Gene Symbol: Zfp503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107250.1 | NP_001100720.1 | 2 | 975 | ||
| GenBank mRNA | BC166993.1 | - | 2 | 990 | ||