siRNA Details
| Gene Symbol: Olr1606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000502.1 | NP_001000502.1 | 1 | 514 | ||
| GenBank mRNA | X89705.1 | - | 1 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1606| Gene Symbol: Olr1606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000502.1 | NP_001000502.1 | 1 | 514 | ||
| GenBank mRNA | X89705.1 | - | 1 | 143 | ||