siRNA Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107253.1 | NP_001100723.1 | 2 | 586 | ||
| XM_006251763.2 | XP_006251825.1 | 3 | 2221 | |||
| XM_006251765.2 | XP_006251827.1 | 2 | 3123 | |||
| GenBank mRNA | BC158799.1 | - | 1 | 532 | ||

Kctd6
Genomic Map