siRNA Details
| Gene Symbol: Abhd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007680.1 | NP_001007681.1 | 4, 5 | 467 | ||
| XM_006251766.2 | XP_006251828.1 | 4, 5 | 1746 | |||
| XM_006251767.2 | XP_006251829.1 | 4, 5 | 1278 | |||
| XM_006251768.2 | XP_006251830.1 | 4, 5 | 443 | |||
| GenBank mRNA | BC083826.1 | - | 4, 5 | 467 | 7112208 | |

Abhd6
Genomic Map