siRNA Details
| Gene Symbol: Samd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107254.1 | NP_001100724.1 | 2 | 613 | ||
| XM_008770515.1 | XP_008768737.1 | 2 | 1451 | |||
| XM_008770516.1 | XP_008768738.1 | 2 | 1450 | |||
| GenBank mRNA | BC168894.1 | - | 2 | 595 | 7453870 | |

Samd4a
Genomic Map