siRNA Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107257.1 | NP_001100727.1 | 1407 | |||
| GenBank mRNA | BC168877.1 | - | 1564 | 7134856 | ||
| FQ212387.1 | - | 1481 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo34| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107257.1 | NP_001100727.1 | 1407 | |||
| GenBank mRNA | BC168877.1 | - | 1564 | 7134856 | ||
| FQ212387.1 | - | 1481 | ||||