siRNA Details
| Gene Symbol: Atg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107258.1 | NP_001100728.1 | 8 | 1100 | ||
| GenBank mRNA | FQ223033.1 | - | 8 | 1068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg14| Gene Symbol: Atg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107258.1 | NP_001100728.1 | 8 | 1100 | ||
| GenBank mRNA | FQ223033.1 | - | 8 | 1068 | ||