siRNA Details
| Gene Symbol: Ttc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013131.1 | NP_001013149.1 | 3 | 254 | ||
| GenBank mRNA | BC091207.1 | - | 3 | 254 | 7381272 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc5| Gene Symbol: Ttc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013131.1 | NP_001013149.1 | 3 | 254 | ||
| GenBank mRNA | BC091207.1 | - | 3 | 254 | 7381272 | |