siRNA Details
| Gene Symbol: Olr1637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000503.1 | NP_001000503.1 | 1 | 896 | ||
| XM_006251869.2 | XP_006251931.1 | 2 | 953 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1637| Gene Symbol: Olr1637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000503.1 | NP_001000503.1 | 1 | 896 | ||
| XM_006251869.2 | XP_006251931.1 | 2 | 953 | |||