siRNA Details
| Gene Symbol: Sall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107262.1 | NP_001100732.1 | 2 | 3059 | ||
| XM_006251884.2 | XP_006251946.1 | 2 | 2594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sall2| Gene Symbol: Sall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107262.1 | NP_001100732.1 | 2 | 3059 | ||
| XM_006251884.2 | XP_006251946.1 | 2 | 2594 | |||