siRNA Details
| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030036.1 | NP_001025207.1 | 2 | 452 | ||
| GenBank mRNA | BC099829.1 | - | 2 | 452 | 7933740 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap5m1| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030036.1 | NP_001025207.1 | 2 | 452 | ||
| GenBank mRNA | BC099829.1 | - | 2 | 452 | 7933740 | |