siRNA Details
| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770650.1 | XP_008768872.1 | 5 | 930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35f4| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770650.1 | XP_008768872.1 | 5 | 930 | ||