siRNA Details
| Gene Symbol: Slc22a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177421.3 | NP_803156.2 | 6 | 1190 | ||
| XM_006251965.1 | XP_006252027.1 | 6 | 1828 | |||
| GenBank mRNA | AB040056.1 | - | 6 | 1197 | ||
| BC099072.1 | - | 6 | 1190 | 7312276 | ||
| FQ212273.1 | - | 6 | 1192 | |||

Slc22a17
Genomic Map