siRNA Details
| Gene Symbol: Zfhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098803.1 | NP_001092273.1 | 9 | 4705 | ||
| XM_008770677.1 | XP_008768899.1 | 8 | 6480 | |||
| XM_008770678.1 | XP_008768900.1 | 7 | 6405 | |||
| GenBank mRNA | AY237644.1 | - | 1 | 18 | ||

Zfhx2
Genomic Map