siRNA Details
Gene Symbol: Ift88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107266.1 | NP_001100736.1 | 9, 10 | 656 | ||
XM_006252066.2 | XP_006252128.1 | 9, 10 | 1638 | |||
XM_006252067.2 | XP_006252129.1 | 10, 11 | 1830 | |||
XM_006252068.2 | XP_006252130.1 | 9, 10 | 939 | |||
XM_006252070.2 | XP_006252132.1 | 9, 10 | 1637 | |||
XM_006252071.2 | XP_006252133.1 | 9, 10 | 1638 |
