siRNA Details
| Gene Symbol: Kif13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213626.1 | NP_998791.1 | 17, 18 | 2034 | ||
| XM_008770753.1 | XP_008768975.1 | 17, 18 | 2256 | |||
| XM_008770754.1 | XP_008768976.1 | 17, 18 | 2256 | |||
| XM_008770755.1 | XP_008768977.1 | 17, 18 | 2256 | |||
| GenBank mRNA | AJ605719.1 | - | 17, 18 | 2034 | ||

Kif13b
Genomic Map