siRNA Details
| Gene Symbol: Scara5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135855.1 | NP_001129327.1 | 8 | 1663 | ||
| XM_006252204.2 | XP_006252266.1 | 8 | 1704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scara5| Gene Symbol: Scara5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135855.1 | NP_001129327.1 | 8 | 1663 | ||
| XM_006252204.2 | XP_006252266.1 | 8 | 1704 | |||