siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107273.1 | NP_001100743.1 | 15 | 2624 | ||
| XM_006252117.2 | XP_006252179.1 | 15 | 2701 | |||
| XM_006252118.2 | XP_006252180.1 | 15 | 2809 | |||
| XM_006252120.2 | XP_006252182.1 | 15 | 2698 | |||
| XM_006252121.2 | XP_006252183.1 | 12 | 2755 | |||
| XM_008770757.1 | XP_008768979.1 | 12 | 2081 | |||

Cdca2
Genomic Map